Cut resistant gloves, HyFlex® 11-318
: Ansell
This uncoated glove provides EN Level 3 cut protection in an 18 gauge liner. Its ultra light fabric, made of Dyneema® Diamond, Spandex, nylon and carbon fibres, creates a sensation of bare hand contact and helps to maintain the hand at a comfortable working temperature.
- Snug/ergonomic fit on the hand thanks to the spandex reinforcement in the tension areas of the hand (knuckles and palm)
- Low linting, silicone- and latex-free and anti-static
- Specific packaging in order to prevent silicone contamination
Ideal for the final assembly of sharp parts, for packaging applications and for quality inspection before painting.
: EN 388:2016 (XX4XB); Cat. II
- Catalog No:
- 112-4722
- 112-4719
- 112-4720
- 112-4721
- 112-4718
- 112-4717
- Model:
- 11-318
- 11-318
- 11-318
- 11-318
- 11-318
- 11-318
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Finish:
- Uncoated
- Uncoated
- Uncoated
- Uncoated
- Uncoated
- Uncoated
- Gauge:
- 18
- 18
- 18
- 18
- 18
- 18
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (XX4XB)
- EN 388:2016 (XX4XB)
- EN 388:2016 (XX4XB)
- EN 388:2016 (XX4XB)
- EN 388:2016 (XX4XB)
- EN 388:2016 (XX4XB)
- Anti-static:
- +
- +
- +
- +
- +
- +