Chemical resistant gloves, AlphaTec® 37-501
: Ansell
These nitrile gloves with cotton flock lining provide excellent resistance to many solvents and oil based chemicals.
- Very good abrasion resistance
- Flexible and comfortable for an excellent dexterity
- Open cuff allows ventilation to keep the hand cool
Ideal for use when handling agrochemicals, meat, fruits and vegetables. Also suitable for chemical processing and preparation, degreasing, maintenance, metal fabrication, tank filling and packaging applications.
: EN ISO 374-1:2016 Type A (AJKLPT), EN ISO 374-5:2016, EN 388:2016 (3101X); Cat. III; AQL 0,65
Food contact approved
Food contact approved
- Catalog No:
- ANSE37-501/10.5
- ANSE37-501/7.5
- ANSE37-501/6.5
- ANSE37-501/9.5
- ANSE37-501/8.5
- Model:
- 37-501
- 37-501
- 37-501
- 37-501
- 37-501
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Finish:
- Raised diamond
- Raised diamond
- Raised diamond
- Raised diamond
- Raised diamond
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- EN norm:
- EN 374 (AJKL), EN 388 (4101)
- EN 374 (AJKL), EN 388 (4101)
- EN 374 (AJKL), EN 388 (4101)
- EN 374 (AJKL), EN 388 (4101)
- EN 374 (AJKL), EN 388 (4101)
- Food contact approved:
- +
- +
- +
- +
- +