Water sampling bottles, glass, wide neck, amber
Leverandør: EP SCIENTIFIC
Sertifikat
Om dette produktet
Type III soda-lime glass, amber, with screw cap
- With PTFE-lined white PP closed top cap (assembled on bottles)
- Certified to meet or exceed EPA Performance Based Specifications for semi-volatile organics, pesticides, PCBs and metals analyses
- Each jar individually barcoded with lot number and unique container number
These bottles are designed for collecting soil, sludge, waste and water samples for a variety of organic and inorganic target analytes where light protection is required. These bottles are processed the same way as the certified jars but without certification documentation, or unprocessed and ready for your own cleaning procedure.
Delivery: Supplied with Certificate of Analysis (CofA) in the case and barcoded with lot number for traceability.
Packaging: Carton sealed with security tape providing tamper evidence. The chipboard divider packaging to cushion and help prevent breakage; overpack required for shipping.
Spesifikasjoner
- Catalog No:
- EPSC141-2500
- EPSC341-0250
- EPSC141-0060
- EPSC341-0060
- EPSC141-0120
- EPSC141-0500
- EPSC341-2500
- EPSC341-0500
- EPSC341-0950
- EPSC141-0250
- EPSC341-0120
- EPSC241-0120
- EPSC341-1250
- EPSC241-0250
- EPSC241-0500
- EPSC241-0950
- EPSC141-0950
- EPSC241-0060
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement