Visual colour kits, DEHA, CHEMets®
Catalog # CHETK-3902
Supplier: CHEMetrics
CAS Number:
Specifications
- Kjemisk metode:PDTS (3-(2-pyridyl)-5,6- bis(4-phenylsulfonic acid)-1,2,4-triazine disodium salt)
- Fp:1 KIT
- Beskrivelse:DEHA
- Område:0, 30, 60, 100, 150, 200, 300, 400 ppb
400, 600, 800, 1000, 1200, 1600, 2000, 2500, 3000 ppb
Specifications
About this item
Ideal for low- to medium-range colorimetric analysis of water quality
CHEMets® test kits utilise self-filling ampoules and vacuum pulls the sample up from a snap cup into the ampoule when the tip is broken off. After tilting the ampoule several times to mix the sample and reagent and waiting for the colour to develop (usually 2 minutes or less), the analyst quantifies the result using the provided comparators.
Delivery: Kit comes in a plastic case and contains everything needed to perform 30 tests: 30 ampoules, low and high range comparators, activator solution, 25 ml sample cup and instructions.