Order Entry
Norway
ContactUsLinkComponent
Pressure relief valves
Pressure relief valves
Catalog # VARIRE7030
Supplier:  VARIAN
Pressure relief valves
Catalog # VARIRE7030
Supplier:  VARIAN
Specifications
  • Description:
    Valve, Rheodyne 7030, three-way switching, stainless steel, 1/16"
  • Pk:
    1
Specifications
About this item
Valves made from stainless steel are two-position with six-port. It is manually operated with three-way switching.