Order Entry
Norway
ContactUsLinkComponent
Mag-Bind® Plant DNA DS 96 kit
Mag-Bind® Plant DNA DS 96 kit
Supplier:  OMEGA BIO-TEK
Certificates
Mag-Bind® Plant DNA DS 96 kit
Supplier:  OMEGA BIO-TEK

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
  • Target molecule:
    DNA
  • Sample size:
    50 mg of wet tissue or 15 mg dry tissue
  • Description:
    Mag-Bind® Plant DNA DS 96 Kit

Specifications