Order Entry
Norway
ContactUsLinkComponent
Gallic acid monohydrate solution in ethylene glycol for conductivity measurement
Gallic acid monohydrate solution in ethylene glycol for conductivity measurement
Catalog # HACH1442326
Supplier:  Hach
CAS Number:  
undefined
Gallic acid monohydrate solution in ethylene glycol for conductivity measurement
Catalog # HACH1442326
Supplier:  Hach
CAS Number:  

Specifications

  • Pack type:
    Dropping bottle, self-contained
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
 :188.14 g/mol
 :GHS07
 :GHS08
CAS Number:5995-86-8
Formula:C₇H₆O₅·1H₂O
MDL Number:MFCD00149098