Spill containment units
Catalog # 121-0184
Supplier: NEW PIG
Specifications
- Capacity:1363 litres l
- Description:PIG® Poly IBC containment unit
- W×D×H:1570×1570×660 mm
- Pk:1
Specifications
About this item
Spill containment units made from LDPE.
- Catch spills if containers leak, drip or crack
- Low profile for easy access to and dispensing from tanks
- Resists UV rays, rust, corrosion and most chemicals
If this product is used with the flammable liquids, please consider the regulations that apply to storage and handling of flammable liquids and the safety of this application, specifically flammable vapours, static discharge and heat sources.