Automated pipetting assistant, VIAFLO ASSIST
613-3481
- Volume:0,5 - 1250 µl
- Description:Pipetting robot, VIAFLO ASSIST for automating multichannel pipettes
- Model:VIAFLO ASSIST
- No. of pipette channels:4, 6, 8, 12 and 16
- Power:70 W
- Power supply:100 - 240 V / 50 - 60 Hz
- Weight of system:10 kg
- W×D×H of system:340×360×400 mm
- Pk:1
VIAFLO ASSIST together with VIAFLO II electronic pipettes form the perfect symbiosis to prevent users from RSI and to significantly increase the reproducibility of prolonged pipetting protocols such as serial dilutions, plate fillings and reagent additions. Simply mount any VIAFLO II electronic pipette onto the VIAFLO ASSIST pipette adaptor, choose a pipetting protocol and press the RUN button. VIAFLO ASSIST will then obtain the protocol via Bluetooth from the electronic pipette and automatically execute the desired application.
- Prevents fatigue and repetitive strain injuries (RSI) caused by extensive repetitive motions
- Ensures accurate tip immersion depth and pipetting angle
- Pipette adapter of VIAFLO ASSIST allows the use of all VIAFLO II multi channel pipettes (8-, 12-, and 16-channel)
- 6-well to 384-well plates can be used; plate sledge accommodates the plates either in portrait or landscape orientation
- Three predefined protocols allow a quick start: serial dilution, repeat dispense and variable dispense
- For more elaborate pipetting routines, custom programs can be created in an easy step by step setup
: VIAFLO ASSIST is supplied with a reservoir sample pack of each size. Multi channel pipettes, tips and reservoirs are not included and need to be ordered separately. Please note that every VIAFLO II pipette, which is used with VIAFLO ASSIST, needs its own Bluetooth module.