3,3'-Diaminobenzidine (DAB) membrane peroxidase substrate (50X)
Catalog # ROCKDAB-10
Supplier: Rockland Immunochemicals
CAS Number:
Specifications
- Pk:10 mL
- Seq:2275
Specifications
About this item
:
:214.27 g/mol
Storage Temperature:Refrigerator
CAS Number:91-95-2
Formula:C₁₂H₁₄N₄
MDL Number:MFCD00007725