Safety needles, HENKE-JECT®
HSWA8300020446
- Ext.Ø:0,80 mm
- Gauge:21
- Type:Non sterile
- Length:120 mm
- Seq:13
- Pk:100
These safety needles are compatible with HENKE-JECT® syringes as well as all others syringes with Luer and Luer lock according to ISO 594.
- Safety mechanism is an integrated part of the needle
- Audible 'click' indicates safety activation
- Single-handed activation
: Conform to EU-Guideline 2010/32/EU and TRBA 250.