Bottle carrier, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Sertifikat
Om dette produktet
HDPE.
- Epoxy coating
- Heavy-gauge steel wire bail handle
- Side ribs and bottom suspend the filled bottle
These bottle carriers are suitable for transport and storage of reagent bottles. The cone-shaped opening allows a delivery tube to be inserted, so liquid does not have to be poured out. The shortened lid allows the bottle to protrude for safer pouring.
Colour: White.