Order Entry
Norway
ContactUsLinkComponent
Glove box, stainless steel, filtered, Protector®
Glove box, stainless steel, filtered, Protector®
Leverandør:  LABCONCO
Sertifikat
undefined
Glove box, stainless steel, filtered, Protector®
Leverandør:  LABCONCO
Spesifikasjoner
  • Catalog No:
  • LBCP5065535
  • LBCP5065537
  • LBCP5065531
  • LBCP5065533
  • Model:
  • Protector® (two glove ports)
  • Protector® (four glove ports)
  • Protector® (two glove ports)
  • Protector® (four glove ports)
  • Observation window (W×H):
  • 914,5×701 mm
  • 914,5×701 mm
  • 914,5×701 mm
  • 914,5×701 mm
  • W×D×H ext.:
  • 1613×787×1161 mm
  • 2766×787×1161 mm
  • 1613×787×1161 mm
  • 2766×787×1161 mm
  • Weight:
  • 295 kg
  • 590 kg
  • 295 kg
  • 590 kg
Spesifikasjoner
Relatert informasjon