Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Furniture
Storage Cabinets
Accessories for drawer cabinets, Sovella
Accessories for drawer cabinets, Sovella
Accessories for drawer cabinets, Sovella
SOVE854850-35
:
TRESTON
Accessories for drawer cabinets, Sovella
SOVE854850-35
:
TRESTON
Description:
Pencil box for drawer units 38 and 45
Pk:
1