Bottles, narrow neck, round, PC, Boston type, with screw caps, Azlon®
Supplier: SciLabware
Certificates
About this item
Polycarbonate, highly transparent.
- Leakproof
- Suitable for repeated sterilisation by autoclaving at +121 °C
- Heavy-duty and durable
Ideal for the preparation and storage of biological media, reagents and aqueous solutions. With linerless polypropylene cap.