Sharps Container
Supplier: USON PLAST
Certificates
About this item
Container, made of PP, yellow.
- Made of environmentally friendly, strong and durable Polypropylene
- Design and manufacturing secure a high degree of safety
- Reversible closure can be safely fixed
- Containers are waterproof when closed
These sharp containers is designed for hospitals and health care institutions to dispose of syringes, scalpels and other types of sharps. Designed to maximise safety whilst still maintaining a barrier to protect the user and prevent spillage during use.
Different lid designs give the possibility to discharge needles, syringes and other types of sharps.
Certifications: Meet the UN requirement no. 3291 regarding packaging approved for transportation of dangerous goods (waste/disposal).