Order Entry
Norway
ContactUsLinkComponent
Muscular figure
Muscular figure
Catalog # BINH1000212
Supplier:  3B Scientific
undefined
Muscular figure
Catalog # BINH1000212
Supplier:  3B Scientific

Specifications

  • Description:
    Muscular figure
  • W×D×H:
    570×250×180 mm
  • Pk:
    1

Specifications

About this item

Mini muscle model representing the superficial musculature of the human form accurately reproduced and detailed in life-like colours in this desktop size version.

  • Over 125 hand numbered and identified structures
  • A third of the life size in two parts
  • Removable chest plate to reveal the internal organs – Right side containing a female mammary gland

Dimensions: 57×25×18 cm

Weight: 2,1 kg

Delivery: Delivered on base.