Blotting papers, Owl™
Supplier: THERMO OWL SCIENTIFIC
Certificates
About this item
Absorb adequate amounts of buffer for semi-dry electroblotting without the loss of mechanical stability with Thermo Scientific™ Owl™ Blotting Paper, high-quality blotting paper ideal for wicking and blotting applications.
Specifications
- Catalog No:
- 700-2190
- OWLAFP-7
- OWLAFP-6
- 700-2192
- 700-2191
- OWLAFP-2
- OWLAFP-3
- Weight:
- 183 g/m²
- 183 g/m²
- 183 g/m²
- 183 g/m²
- 183 g/m²
- 183 g/m²
- 183 g/m²
- Thickness:
- 0,83 mm
- 0,83 mm
- 0,83 mm
- 0,83 mm
- 0,83 mm
- 0,83 mm
- 0,83 mm