Order Entry
Norway
ContactUsLinkComponent
Air gel drying kits and supplies, Owl™
  700-2101
undefined
Air gel drying kits and supplies, Owl™
  700-2101
  • Description:
    Precut cellophane, 25×25 cm
  • Seq:
    06
  • Pk:
    50
Preserve protein gels easily and economically while eliminating distortion problems common in other drying procedures with these gel-drying kits.