Hexnut fittings
Supplier: Trajan Scientific and Medical
Certificates
About this item
Stainless steel 10-32 thread fittings use a non-swaging PEEK replaceable ferrule, for use with GLT™ columns.
- Inert contact surfaces make them biocompatible
- Ideal for applications where corrosive solvents are being used
- Maximum pressure rating of 344,74 bar (5000 psi)