Industrial gloves, 380
: Showa
These seamless knitted nylon gloves feature a microporous nitrile coating on the palm which protects the hand and provides an optimal grip.
- Palm coating
- Foamed embossed finish for optimal grip in dry and slightly oily environments
- Elasticated cuff, anatomical shape
- Lightweight and low-linting
- Robust, great dexterity and good tearing resistance
- Breathable back reduces perspiration
Ideal for light assembly tasks in construction industries and in the maritime sector.
: EN 388:2016 (3121X); Cat. II
- Catalog No:
- 112-5824
- 112-5821
- 112-5823
- 112-5822
- Model:
- 380
- 380
- 380
- 380
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Nylon
- Nylon
- Nylon
- Nylon
- Finish:
- Embossed palm and fingertips coating
- Embossed palm and fingertips coating
- Embossed palm and fingertips coating
- Embossed palm and fingertips coating
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (3121X)
- EN 388:2016 (3121X)
- EN 388:2016 (3121X)
- EN 388:2016 (3121X)