Order Entry
Norway
ContactUsLinkComponent
Tape dispensers
Tape dispensers
  MICRP-52W
 :  MICRONOVA
Tape dispensers
  MICRP-52W
 :  MICRONOVA

 

  • Description:
    Tape dispenser for 5,08 cm wide tapes, with weighted base
  • Seq:
    02
  • Pk:
    1

 

 

This tape dispenser is a combination of coated steel and plastic. Designed for use with Micronova tapes of up to 7,62 cm width.

  • Available with or without weighted base
  • Available in 5,08 cm and 15,24 cm width capacities