Cut resistant gloves, Camapur® Cut 627
Supplier: KCL by Honeywell
Certificates
About this item
This glove made from HPPE fibres combines good sensitivity, mechanical resistance and high cut-resistance. The thin PU coating offers good sensitivity and prevents sweating.
- Excellent grip, even when handling slightly oiled, wet and slippery items
- Back of hand without coating for improved breathability
- Dark grey colour for reduced susceptibility to dirt
- Size of the glove is distinguishable at the knitted cuff through different colours
Ideal for assembly work with medium cutting risk, in sheet metal processing and the processing of plastic profiles.
Thickness: 1,2 ± 0,1 mm
Colour: grey/black
EN 388 (4341); Cat II
Certifications: EN 388:2016 (4X41B); Cat. II
Specifications
- Catalog No:
- 112-2978
- KCHL627.12
- KCHL627.6
- KCHL627.7
- 112-2977
- 112-2976
- KCHL627.11
- Model:
- Cut 627
- Cut 627
- Cut 627
- Cut 627
- Cut 627
- Cut 627
- Cut 627
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- HPPE
- HPPE
- HPPE
- HPPE
- HPPE
- HPPE
- HPPE
- Finish:
- Coating on palm and fingers
- Coating on palm and fingers
- Coating on palm and fingers
- Coating on palm and fingers
- Coating on palm and fingers
- Coating on palm and fingers
- Coating on palm and fingers
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4X41B)
- EN 388:2016 (4X41B)
- EN 388:2016 (4X41B)
- EN 388:2016 (4X41B)
- EN 388:2016 (4X41B)
- EN 388:2016 (4X41B)
- EN 388:2016 (4X41B)