Order Entry
Norway
ContactUsLinkComponent
Accessories standard pH electrodes
Accessories standard pH electrodes
Catalog # GRSG601382
Supplier:  Greisinger
undefined
Accessories standard pH electrodes
Catalog # GRSG601382
Supplier:  Greisinger
Specifications
  • Description:
    Adapter for the plug-in of electrodes with BNC-plugs to devices with Cinch-sockets
  • Pk:
    1