ESD occupational shoes, sandals, Nature, 4080/4090
Supplier: Abeba
Certificates
About this item
ESD sandals made from smooth leather with or without adjustable heel strap and a rubber sole (resistant to fuels, oils, acids and bases).
- Skin friendly, leather coated, cork integrated insole with toe gripper
- Sanitized® treatment reduces odour and bacterial growth
- Slip resistant special rubber sole with shock-absorber heel
- Adjustable instep area with Velcro® fastening
Colour: White
Colour: White
Certifications: EN 61340 (ESD); TÜV GS DIN 4843/100 § 3.1v.8/93, EN ISO 20347:2012; OB, FO, A, SRC