Occupational shoes, clogs, The Original, 5080/5081
Supplier: Abeba
Certificates
About this item
Anti-static clogs made from smooth leather or perforated leather upper and a sole with cross/circle profile resistant to animal fats, oil and fuel.
- Heel strap is foldable and adjustable
- Leather lining
- Sanitized® treatment reduces odour and bacterial growth
Colour: White leather, green sole
Colour: White leather, green sole
Certifications: TÜV GS-tested; EN ISO 20347:2012; OB, SRC