Disarticulated human skeleton
BINH1000026
- Weight:6 kg
- Description:Disarticulated human skeleton
- W×D×H:485×270×425 mm
- Pk:1
This disarticulated skeleton is hand-painted and numbered to show muscle origins in red and muscle insertions in blue on the left side. Bones and bony structures such as fissures, foramina and processes are hand-numbered on the right side. The skull is detachable into three parts for detailed anatomical study. The muscle disarticulated full skeleton features:
- One hand and foot on wire
- One set of arms and leg loosely
- Multilingual product manual to identify over 600 structures
: Supplied in a sturdy partitioned cardboard storage box.