General purpose gloves, HyFlex® 11-818
Supplier: Ansell
Certificates
About this item
This knitted spandex/nylon liner features a foam nitrile coating on the palm. Under abrasive conditions the ultra-thin FORTIX™ nitrile foam coating lasts up to three times longer than comparable coatings, extending glove life and reducing total cost of ownership.
- FORTIX™ coating provides enhanced dry grip and is breathable, for cooler, drier hands and less sweat
- Seamless 18-gauge nylon-spandex liner has a barehand feel, offering breathability, comfort and enhanced fingertip sensitivity for precision tasks
- Ansell ZONZ® Comfort Fit technology matches natural hand contours for improved support and freedom of movement
- Ultra-lightweight, abrasion-resistant
- Silicone-free material means no transfer of silicone contaminants to metal parts prior to painting
- Oeko-Tex® certified
Ideal for the assembly of dry parts, fine mounting of small nuts and bolts, handling of small/fine parts, mounting and dismounting parts, screwing and unscrewing and ultra-fine mechanical applications.
Certifications: EN 388:2016 (3121A); Cat. II
OEKO-Tex® Standard 100
OEKO-Tex® Standard 100
Specifications
- Catalog No:
- 112-5134
- 112-5133
- 112-5132
- 112-5131
- 112-5136
- 112-5135
- Model:
- 11-818
- 11-818
- 11-818
- 11-818
- 11-818
- 11-818
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Spandex/nylon
- Spandex/nylon
- Spandex/nylon
- Spandex/nylon
- Spandex/nylon
- Spandex/nylon
- Finish:
- Foam coating on palm
- Foam coating on palm
- Foam coating on palm
- Foam coating on palm
- Foam coating on palm
- Foam coating on palm
- Gauge:
- 18
- 18
- 18
- 18
- 18
- 18
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (3121A)
- EN 388:2016 (3121A)
- EN 388:2016 (3121A)
- EN 388:2016 (3121A)
- EN 388:2016 (3121A)
- EN 388:2016 (3121A)