Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Methylthio)benzaldehyde
2-(Methylthio)benzaldehyde ≥97.0% (by GC)
2-(Methylthio)benzaldehyde ≥97.0% (by GC)
:
TCI
2-(Methylthio)benzaldehyde ≥97.0% (by GC)
:
TCI
:
7022-45-9
:
:
C₈H₈OS
:
MFCD00196822
:
152.2135 g/mol
:
149°C
:
Refrigerator
Environmentally Preferable:
Appearance:
Light orange to Yellow to Green clear liquid
Purity (GC):
min. 97.0 %