
Benzo-12-crown-4 (Benzo-1,4,7,10-tetraoxacyclododecane) ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:14174-08-4
:
Formula:C₁₂H₁₆O₄
MDL Number:MFCD00059934
:224.26 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:45.0 to 49.0 deg-C