Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Ordreinnføring
Ordrestatus
Din konto
Handlevogn
Norway
Ordreinnføring
Ordrestatus
Din konto
Handlevogn
ContactUsLinkComponent
Hjem
Coomassie brilliant blue G-250
Coomassie brilliant blue G-250
Coomassie brilliant blue G-250
Coomassie brilliant blue G-250
Supplier:
Cayman Chemical
Certificates
Expand image
Coomassie brilliant blue G-250
Supplier:
Cayman Chemical
About this item
CAS Number:
6104-58-1
:
Formula:
C₄₇H₄₈N₃NaO₇S₂
MDL Number:
MFCD00078482
:
854.04 g/mol
Density:
>1 g/cm³ (20 °C)
Storage Temperature:
Ambient
EINECS:
228-058-4