Order Entry
Norway
ContactUsLinkComponent
Luff-Schoorl's reagent 25 g/L volumetric solution, Fluka™
Luff-Schoorl's reagent 25 g/L volumetric solution, Fluka™
Certificates
Luff-Schoorl's reagent 25 g/L volumetric solution, Fluka™
Certificates

Specification Test Results

  • Assay of CuSO₄*5H₂O:
    24.9 - 25.1 g/l
  • pH:
    9.3 - 9.4

Specification Test Results

Related Information