Pyridine-2,4-dicarboxylic acid
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:499-80-9
:MFCD00006296
Formula:C₇H₅NO₄
MDL Number:MFCD00149413
:167.12 g/mol
Boiling Pt:357 °C (1013 hPa)
Melting Pt:246…248 °C
Density:0.942 g/cm³ (25 °C)
EINECS:207-892-2