6-Propyl-2-thiouracil
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:51-52-5
:
Formula:C₇H₁₀N₂OS
MDL Number:MFCD00006041
:170.24 g/mol
Merck Index:12,08054
Boiling Pt:448 °C (1013 hPa)
Melting Pt:215…218 °C
EINECS:200-103-2
:GHS07