4-(Methylsulphonyl)benzonitrile 95%
ACRO424890025
:
- Pakketype:Glass bottle
- Fp:2,5 g
- Seq:3122
- Appearance:White to orange to brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:137 to 142 °C
- GC:≥94 %
:
:181.2100 g/mol
:137…142 °C
:GHS07
:Romtemperatur
:22821-76-7
:C₈H₇NO₂S
:MFCD00216489