Order Entry
Norway
ContactUsLinkComponent
 
 
4-(tert-Butyldimethylsilyloxy)phenylboronic acid (contains varying amounts of Anhydride)
 :  TCI
 
undefined
 
4-(tert-Butyldimethylsilyloxy)phenylboronic acid (contains varying amounts of Anhydride)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    97.0 to 108.0 %