Pilocarpine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:54-71-7
:
Formula:C₁₁H₁₇ClN₂O₂
MDL Number:MFCD00012722
Merck Index:14,07424
EINECS:200-212-5
UN:1544
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:201.0 to 205.0 deg-C
- Specific rotation [a]20/D:+89.0 to +94.0 deg (C=2, H2O)