Order Entry
Norway
ContactUsLinkComponent
Pilocarpine hydrochloride
Pilocarpine hydrochloride
Katalog # MOLE26311033-5G
Supplier:  Molekula
CAS Number:  
Pilocarpine hydrochloride
Katalog # MOLE26311033-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
EINECS:200-212-5
CAS Number:54-71-7
Formula:C₁₁H₁₇ClN₂O₂
MDL Number:MFCD00012722