Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Fmoc-L-valine
N-Fmoc-L-valine
N-Fmoc-L-valine
N-Fmoc-L-valine
Supplier:
Novabiochem (Part of Merck)
Certificates
Expand image
N-Fmoc-L-valine
Supplier:
Novabiochem (Part of Merck)
Certificates
About this item
CAS Number:
68858-20-8
:
Formula:
C₂₀H₂₁NO₄
MDL Number:
MFCD00037124
:
339.4 g/mol
EINECS:
272-515-0
Related Information
Documentation
Certificates
Search by Lot Number
Documentation