Gold zinc alloy (90:10 w%) ≥99.95% (metals basis), slug 3.175mm (0.125in) dia x 3.175mm (0.125in) length
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
MDL Number:MFCD02092916
Storage Temperature:Ambient
Related Information
Frequently Bought Together