Order Entry
Norway
ContactUsLinkComponent
Accessories for EBA 20 centrifuge
Accessories for EBA 20 centrifuge
  HETT6305
 :  Hettich
   Low Manufacturing Impact
Accessories for EBA 20 centrifuge
  HETT6305
 :  Hettich
   Low Manufacturing Impact

 

  • Description:
    Adapter, PP, for 1×4 ml round-bottom tube (Ø×L: 11,5×67,5 mm)
  • Seq:
    02
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
  • Pk:
    1