Order Entry
Norway
ContactUsLinkComponent
Isonicotinaldehyde ≥96.0% (by GC)
Irritant
Isonicotinaldehyde ≥96.0% (by GC)
Catalog # TCIAI0143-25ML
Supplier:  TCI
CAS Number:  
undefined
Irritant
Isonicotinaldehyde ≥96.0% (by GC)
Catalog # TCIAI0143-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Purity (GC):
    min. 96.0 %

Specification Test Results

About this item

 :
 :107.11 g/mol
Boiling Pt:187°C
Flash Pt:75°C
EINECS:212-832-3
 :GHS07
CAS Number:872-85-5
Formula:C₆H₅NO
MDL Number:MFCD00006425