Ammonium sulphide 1% in aqueous solution
Katalog # LBSE790838
Supplier: LABSERVICE
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
:68.14 g/mol
EINECS:235-223-4
UN:2683
ADR:8,II
CAS Number:12135-76-1
Formula:H₈N₂S
MDL Number:MFCD00010892