
2-Mercapto-5-methyl-1,3,4-thiadiazole ≥98.0% (by titrimetric analysis)
Catalog # TCIAM0976-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:184.0 to 188.0 deg-C
Specification Test Results
About this item
:
:132.21 g/mol
EINECS:249-667-1
:GHS07
CAS Number:29490-19-5
Formula:C₃H₄N₂S₂
MDL Number:MFCD00038349