Order Entry
Norway
ContactUsLinkComponent
(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥97.0% (by titrimetric analysis)
(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥97.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥97.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Optical purity (LC):
    min. 98.0 ee%
  • Purity (Neutralization titration):
    min. 97.0 %
  • Specific rotation [a]20/D:
    +580.0 to +605.0 deg (C=1, MeOH)
  • Solubility in hot Methanol:
    almost transparency

Specification Test Results

Frequently Bought Together