(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate
Catalog # MOLE76943966-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:348.289 g/mol
CAS Number:35193-64-7
Formula:C₂₀H₁₃O₄P
MDL Number:MFCD00010045