Order Entry
Norway
ContactUsLinkComponent
Phenol : chloroform : iso-amyl alcohol (125:24:1) (v/v/v) ≥99.0%, Biotechnology Grade
Phenol : chloroform : iso-amyl alcohol (125:24:1) (v/v/v) ≥99.0%, Biotechnology Grade
Catalog # E277-400ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
Phenol : chloroform : iso-amyl alcohol (125:24:1) (v/v/v) ≥99.0%, Biotechnology Grade
Catalog # E277-400ML
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    400 mL
  • Seq:
    2665

Specifications

Specification Test Results

  • Abs.@ 330 nm:
    0.2
  • Abs.@ 405 nm :
    0.1
  • Abs.@ 510 nm :
    0.1
  • pH @ 25 °C:
    4.3 – 4.7
  • Purity:
    99.0

Specification Test Results

About this item

 :
UN:2810
ADR:6.1,II
 :GHS06
 :GHS05
 :GHS08
Storage Temperature:Ambient
CAS Number:136112-00-0
MDL Number:MFCD00133763