Citric acid, anhydrous ≥99.5%, powder, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:77-92-9
:
Formula:HOC(COOH)(CH₂COOH)₂
MDL Number:MFCD00011669
:192.13 g/mol
Merck Index:13,02350
Boiling Pt:310 °C (1013 hPa)
Melting Pt:149…151 °C
Density:1.665 g/cm³ (18 °C. OECD 109)
Storage Temperature:Ambient
EINECS:201-069-1
REACH:01-2119457026-42
:GHS07
Specification Test Results
- Assay:Min. 99.5 %
- Heavy metals (as Pb):Max. 10 ppm
- Water:Max. 0.2 %
- Pb (Lead):Max. 5 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC84842.290 spec_VWRC84842.290 spec_VWRC84842.360 spec_VWRC84842.360