Face shields, ArcPro™ anti electric arc systems
Supplier: CENTURION SAFETY PRODUCTS
Certificates
About this item
These systems have been designed to protect against arc flash risks posed by electrical faults.
- Clear PC screen protects against short circuit arcs
- Green PC screens also feature anti-fog coating
- Face screens supplied with chin guard made of fibre reinforced nylon
The face screens are available without or in complete kits with helmet.
Certifications: EN 166 1 B 8 9 (EN 170 2-1,2); Arc exposure test according to: GS-ET-29:2010, Class 2 (423 kJ/m²)