Order Entry
Norway
ContactUsLinkComponent
Young common viper
Young common viper
Catalog # SOMMZOS1036
Supplier:  SOMMER MARCUS
undefined
Young common viper
Catalog # SOMMZOS1036
Supplier:  SOMMER MARCUS

Specifications

  • Vekt:
    1,1 kg
  • Beskrivelse:
    Young common viper
  • Materiale:
    SOMSO-Plast®
  • B×D×H:
    340×180×180 mm
  • Fp:
    1

Specifications

About this item

Vipera. Including label and printed description.

Packaging: On a green base under a transparent box.